Learn More
Cowie Technology™ PTFE Straight Through Stopcocks
Cowie Technology™ PTFE Straight Through Stopcocks feature a pure PTFE body and plug and combine all the desirable properties of the Ultimate Laboratory Stopcock.
Brand: Cowie Technology™ 016.003
Weitere Details : Weight : 0.04600kg
Beschreibung
- Medium vacuum pressure performance
- Unbreakable
- Chemically inert
- Grease free
- Supplied with borosilicate glass sidearms
Spezifikation
Straight Through | |
Straight Through Stopcock | |
Autoclavable | |
1 Each | |
PTFE |
3 mm | |
PTFE | |
8 mm | |
Borosilicate glass sidearms |