missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate, Extra Pure, SLR, Fisher Chemical™
39.95 EUR - 138.00 EUR
Chemical Identifiers
CAS | 7758-05-6 |
---|---|
Molecular Formula | IKO3 |
Molecular Weight (g/mol) | 214.00 |
MDL Number | 11406 |
InChI Key | JLKDVMWYMMLWTI-UHFFFAOYSA-M |
Synonym | potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat |
PubChem CID | 23665710 |
IUPAC Name | potassium;iodate |
SMILES | [O-]I(=O)=O.[K+] |
Description
Chemical Identifiers
7758-05-6 | |
214.00 | |
JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
23665710 | |
[O-]I(=O)=O.[K+] |
IKO3 | |
11406 | |
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
potassium;iodate |
Specifications
560°C | |
White | |
IKO3 | |
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
[O-]I(=O)=O.[K+] | |
214.00 | |
214g/mol |
7758-05-6 | |
Powder/Solid | |
11406 | |
JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
potassium;iodate | |
23665710 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium iodate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.