Learn More
Macherey-Nagel™ Chromafil™ Xtra PTFE Syringe Filters
Filter nonpolar liquids and gases. Chromafil™ Xtra PTFE Syringe Filters with polytetrafluoroethylene hydrophobic membrane.
129.00 EUR - 515.00 EUR
Specifications
Material (Membrane) | PTFE |
---|---|
Filtration Area | ∼4.9 cm2 |
Housing Material | Polypropylene (PP) |
Type | Syringe Filter |
Volume (Metric) | ∼80 μL |
Product Code | Brand | Pore Size | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Quantity | Price | Quantity & Availability | ||||
10787548
|
Macherey-Nagel™
729207 |
0.20 μm | 100 Pack |
129.00 EUR
Pack of 100 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
10762454
|
Macherey-Nagel™
729205 |
0.45 μm | 100 Pack |
129.00 EUR
Pack of 100 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
10045793
|
Macherey-Nagel™
729205.400 |
0.45 μm | 400Pack |
515.00 EUR
Pack of 400 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
Description
- Hydrophobic membrane
- For nonpolar liquids and gases
- Very resistant towards all kinds of solvents as well as acids and bases
- Flushing with alcohol, followed by water, makes the originally hydrophobic membrane more hydrophilic
Specifications
PTFE | |
Polypropylene (PP) | |
∼80 μL |
∼4.9 cm2 | |
Syringe Filter | |
25 mm |