Learn More
Cole-Parmer™ Polysulfone All-Plastic Quick-Disconnect Sealing Cap
Brand:  Cole-Parmer™ 31311-74
Product Code. 12393770
  • 49.70 EUR / Pack of 5
Estimated Shipment: 18-11-2024
Specifications
Quick-Disconnect Sealing Cap
Fits 0.37 in. Tubing I.D.
5 Pack
Polysulfone
-40°C
148°C