Learn More
Buerkle™ Polystyrene Volumetric Spoons
The capacity is embossed into the spoon handle. Available as a set with 8 volumetric spoons or separate in bags of 100 volumetric spoons.
44.63 EUR
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
13427888
|
Buerkle™
5334-0005 |
0.5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13447888
|
Buerkle™
5334-0010 |
1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Polystyrene, blue
- Round shape, easy emptying
Specifications
Volumetric Spoon | |
Polystyrene | |
Blue |