Learn More
Bel-Art™ SP Scienceware™ Polypropylene Water Bath Rack
Submersible, Sinks without Adding Weight
Brand: Bel-Art™ F18863-0001
This item is not returnable.
View return policy
Description
- Durable polypropylene rack that sinks to the bottom of a water bath and remains upright when agitated
- Steam autoclavable at 121°C (250°F)
- Dimensions: 8.375L x 4.312 in.W (213 x 110mm)
Specifications
Autoclavable | |
Water Bath Rack | |
72 Test Tubes | |
21.3 x 11 x 11.1 cm | |
11 cm |
White | |
10 to 13 mm Test Tubes | |
Polypropylene | |
21.3 cm | |
11.1 cm |