Learn More
Masterflex™ Bulkhead Union Compression Fitting, Polypropylene
Connect rigid tubing systems with these bulkhead compression fittings. Polypropylene Bulkhead Union Compression Fittings form a tight seal and withstand higher pressures than barbed fittings.
Brand: Masterflex™ 06390-10
This item is not returnable.
View return policy
Description
Assemble fittings to tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.- Excellent chemical resistance
- With plastic grippers
- Rated to 220 psi maximum at 25 °C
- Usable from 0 to 100 °C
- Sterilization: Ethylene Oxide (ETO)
Specifications
Compression Fitting | |
1/4 in. tubing O.D. | |
10 Pack | |
220 psi |
Polypropylene | |
0°C | |
7/32 in. orifice; 2-7/32 x 5/8 in. | |
100°C |