Learn More
Menshen™ Polypropylene Screw Caps Wide Neck, Blue
Use Kautex™ Polypropylene Screw Caps Wide Neck, Blue to top off wide neck bottles used in industry or the laboratory
40.00 EUR
Specifications
Closure Material | Polypropylene |
---|---|
Closure Type | Screw Cap |
Product Type | Screw Cap |
Color | Blue |
Material | Polypropylene |
Product Code | Brand | Certifications/Compliance | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Certifications/Compliance | Price | Quantity & Availability | |||||
10412022
|
Menshen™
2000073199 |
DIN 54 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Blue
- For 500-1000ml bottle
- PP
Specifications
Polypropylene | |
Screw Cap | |
Polypropylene | |
1 Each | |
Polypropylene |
Screw Cap | |
Blue | |
1 Each | |
Blue | |
Blue |