Learn More
Fisherbrand™ Polyethylene Sample Bags
For use in the laboratory to carry samples into a sterile environment.
44.95 EUR - 230.00 EUR
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11934385
|
Fisherbrand™
|
Capacity: 1600mL; Closure Type: Round Wire Closure |
195.00 EUR
Pack of 250 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11984385
|
Fisherbrand™
|
Capacity: 1600mL; Closure Type: Flat Wire Closure |
230.00 EUR
Pack of 250 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
11974395
|
Fisherbrand™
|
Capacity: 720mL; Round wire closure |
168.00 EUR
Pack of 500 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
11944405
|
Fisherbrand™
|
Capacity: 60mL; Round wire closure |
44.95 EUR
Pack of 500 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
11984395
|
Fisherbrand™
|
Capacity: 540mL; Closure Type: Flat Wire Closure |
103.00 EUR
Pack of 500 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
11954405
|
Fisherbrand™
|
Capacity: 1.26L; Round wire closure |
195.00 EUR
Pack of 500 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
11974405
|
Fisherbrand™
|
Capacity: 1.26L; Closure Type: Flat Wire Closure |
195.00 EUR
Pack of 500 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||||
Description
- Secure, contaminant free bags to ensure dependable analysis results
- Made from heavy gauge, virgin polyethylene that meets FDA specifications
- Can accommodate solid, semi solid and liquid materials
- Easy to use, insert sample and seal by twirling the bag away outward four times and bending wire ends inward
- Economical and efficient way to collect, contain and carry samples