Learn More
Masterflex™ Polycarbonate Female Luer Fitting
19.00 EUR - 44.80 EUR
Specifications
Product Type | Female Luer Fitting |
---|---|
Material | Polycarbonate |
Packaging Quantity | 25 Pack |
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
13538810
|
Masterflex™
45501-08 |
Fits Female luer x 3/16 in. hose barb adapter |
23.05 EUR
Pack of 25 |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
13477728
|
Masterflex™
45501-22 |
Fits Female luer x female luer adapter |
21.05 EUR
Pack of 25 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15819911
|
Masterflex™
45501-00 |
Fits Female luer x 4mm (5/32 in.) hose barb adapter |
20.40 EUR
Pack of 25 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
13551060
|
Masterflex™
45501-06 |
Fits Female luer x 4mm (5/32 in.) hose barb adapter |
19.00 EUR
Pack of 25 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
13518810
|
Masterflex™
45501-20 |
Fits Female luer x 1/4 in. hose barb adapter |
20.10 EUR
Pack of 25 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
13487728
|
Masterflex™
45501-56 |
Fits Female x Female x Female |
44.80 EUR
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Specifications
Female Luer Fitting | |
Polycarbonate | |
25 Pack |