Learn More
WLD Tec™ Platinum Inoculating Loop for Microbiology
Brand:  WLD Tec™ 6.000.383
Product Code. 10669652
  • 136.00 EUR / Each
Only null left
Estimated Shipment: 28-07-2025
Specifications
6 mm
Platinum
Inoculating Loop
1 Ea.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WLD Tec™ Platinum Inoculating Loop for Microbiology > Diameter: 6mm