missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-TDP43 (Ser403, Ser404) Mouse anti-Human, Mouse, Clone: 6B11B12, Proteintech
Mouse Monoclonal Antibody
130.00 EUR - 350.00 EUR
Specifications
Antigen | Phospho-TDP43 (Ser403, Ser404) |
---|---|
Clone | 6B11B12 |
Concentration | 1.14 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897625
|
Proteintech
66079-1-IG-20UL |
20 μL |
130.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887625
|
Proteintech
66079-1-IG-100UL |
100 μL |
350.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Product No. 66079-1-Ig is a mouse monoclonal antibody recognizing TDP-43 only when phosphorylated at 403/404. Immunohistochemical analyses using this antibody only stain the insoluble inclusions in pathologic tissues without normal diffuse nuclear stainin
Various biochemical, physiological and behavioural processes display circadian rhythms controlled by an internal biological clock. The central "gears" driving this clock appear to be composed of an autoregulatory transcription/posttranslation-based feedback loop. Cryptochrome 1 (CRY1) and 2 (CRY2) are DNA-binding flavoproteins that bear some homology to blue-light receptors and photolyases. In Drosophila, CRY is a photoreceptor for the circadian clock where it binds to the clock component TIM in a light-dependent fashion and blocks its function. Mammalian CRY1 and CRY2 function via light-independent interactions with circadian genes CLOCK and BMAL1, as well as with PER1, PER2, and TIM. They seem to act as light-independent components of the circadian clock and likely regulate Per1 transcriptional cycling via interactions with both the activator and its feedback inhibitors. Mutant mice not expressing the Cry1 or Cry2 protein display accelerated and delayed periodicity of locomotor activity, respectively. It appears that the combination of both proteins working together is essential to synchronize the organism to circadian phases. A critical balance between Cry1 and Cry2 is required for proper clock function; in complete darkness, double-mutant mice present with instantaneous arrhythmicity, indicating the absence of an internal circadian clock.Specifications
Phospho-TDP43 (Ser403, Ser404) | |
1.14 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ALS10, TAR DNA binding protein, TAR DNA binding protein 43, TARDBP, TDP 43, TDP43 | |
Tardbp | |
IgG2a | |
Protein A | |
Antibody |
6B11B12 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Human | |
Q13148, Q921F2 | |
230908, 23435 | |
Peptide | |
Primary | |
-20°C | |
TARDBP |