Learn More
3M™ PELTOR™ Kids Ear Muffs
Click to view available options
Color:
Black, Pink
Green
Specifications
Color | Black, Pink |
For Use With (Application) | Designed For Younger Wearers, Including Children, To Protect Against Moderate Levels of Noise in Everyday Situation Such as Social and Leisure Activities. |
Noise Reduction Rating (NRR) | 27 dB |
Plug Shape | Oval |
Product Type | Kid Ear Muffs |
Style | Headband |