Learn More
WTW™ OxiTop™ IS BOD Self Check Measurement System
A complete respirometric measuring system for BOD self-check with undiluted samples featuring ranges of up to 4,000mg/L BOD.
609.81 EUR - 1721.47 EUR
Specifications
Display Type | LED |
---|---|
Range | 0 to 4000 mg/L BSB |
For Use With (Application) | Respirometric self-check measurement |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11751243
|
WTW™
208817 |
6 OxiTop™ measuring heads (3 yellow, 3 green), 6 PF600 sample bottles, 6 GK 600 Rubber sleeves and 6 RST 600 magnetic stirrers |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11741243
|
WTW™
208819 |
2 OxiTop™ measuring heads (yellow, green), 2 PF600 sample bottles, 2 GK 600 Rubber sleeves and 2 RST 600 magnetic stirrers |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Easy handling without dilution series
- Data security by built-in measured value memory
- Suitable for immediate use
- Based on pressure management in a closed system - the microorganisms in the sample consume the oxygen and generate CO2 in the process
- OxiTop™ heads have an AutoTemp function
- Automatic saving of 5 measured values, manual reading of values after 5 days have elapsed
Specifications
LED | |
0 to 4000 mg/L BSB | |
Respirometric self-check measurement |