Learn More
Thermo Scientific™ Owl™ A1 Large Gel Electrophoresis System Standard Comb
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs. For use with the Fisherbrand™ Owl™ A1 Large Gel Electrophoresis Systems.
Brand: Thermo Scientific™ A1-RL-25C
48.80 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 0.22000kg
Description
Eliminate comb assembly with this assortment of heavy-duty, one-piece Thermo Scientific™ Owl™ A1 Large Gel Electrophoresis System Combs, for use with Owl A1 Large Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.Specifications
Rapid load comb | |
Owl | |
25 (2X) |
Owl A1 Large Gel Electrophoresis System | |
Comb | |
1.0 mm |