Learn More
Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized, Thermo Scientific Chemicals
48.30 EUR - 507.00 EUR
Specifications
Chemical Name or Material | Osmium tetroxide |
---|---|
CAS | 20816-12-0, 75-65-0 |
Color | White to Yellow |
Physical Form | Crystals, Fused Mass or Solution |
Molecular Formula | O4Os |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10207073
|
Thermo Scientific Acros
197450010 |
1 mL | Glass bottle |
48.30 EUR
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10256970
|
Thermo Scientific Acros
197450050 |
5 mL | Glass bottle |
152.00 EUR
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10003433
|
Thermo Scientific Acros
197450250 |
25 mL | Glass bottle |
507.00 EUR
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Osmium tetroxide | |
White to Yellow | |
O4Os | |
MFCD00011150 | |
4°C | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
O=[Os](=O)(=O)=O | |
254.23 | |
254.2 | |
Glass bottle | |
0.8110g/mL |
20816-12-0, 75-65-0 | |
Crystals, Fused Mass or Solution | |
OsO4 | |
01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236 | |
15, 6990 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
tetraoxoosmium | |
30318 | |
2.5 wt% Min. | |
0.811 |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause allergy or asthma symptoms o
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 244-058-7
missing translation for 'rtecsNumber' : RN1140000
missing translation for 'tsca' : TSCA
- Osmium tetroxide, 2.5 wt% solution in t-butanol, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- H301-Toxic if swallowed.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.