Learn More
Diba Omnifit™ Replacement Endpieces for EZ Columns
Select from fixed or adjustable Omnifit™ Replacement Endpieces for EZ Columns
Brand: Diba 006EFE-15
Additional Details : Weight : 0.11000kg
Specifications
15 mm | |
Fixed | |
Omnifit™ EZ Columns |
Omnifit™ EZ EZ Fixed Column Endpiece, 15mm; 1/ea | |
Liquid Chromatography Column | |
PTFE |