Learn More
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Adjustable Endpieces
Omnifit™ EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces
Brand:  Diba 006EZS-35-15-AA
Additional Details : Weight : 0.79000kg
 View more versions of this product
Product Code. 15805494
  • POA / Each
Estimated Shipment: 18-10-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
  • PEEK™ column threads offer outstanding durability
  • Ergonomically designed adjustable endpiece offers quick and easy height adjustment
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Specifications
Specifications
35 mm
Omnifit™ EZ SolventPlus™ EZ SolventPlus Chromatography Column w/ 2 Adjustable Endpieces, 35 x 150mm; 1/ea
150 psi
2.3 to 115.5 mL
Borosilicate Glass, PTFE, Chemraz™
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting.
4 to 40°C
Liquid Chromatography Column
10 and 30 μm
150 mm
40°C