Learn More
asecos™ Oil Sorbents Cushions
The Oil Sorbents Cusions are water repellant for the safe absorption of oils, petrol, diesel, etc.
89.04 EUR - 129.32 EUR
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
17856436
|
asecos™
9933 |
250.00 mm | 250.00 mm |
129.32 EUR
Pack of 20 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
17896546
|
asecos™
9932 |
460.00 mm | 460.00 mm |
89.04 EUR
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
Description
- Very strong durability
- Suitable for heavy and long-lasting use
- Tightly woven fibres, high abrasion resistance and low-lint surface
- Outstanding absorption performance
- Large absorption capacity (up to 16 time of own weight)
- Light for smaller leakages and prevention
- Heavy for larger leakages or absorption of puddles
Specifications
White | |
Heavy-duty Cloth | |
Oil Sorbent Pillow |