Learn More
Welch Ilmvac™ Oil Mist Filter
Eliminate the emission of oil mist. Welch Ilmvac™ Oil Mist Filters feature transparent polyamide housing that enables an easy check of the degree of contamination or saturation of the filter.
Brand:  Welch Ilmvac™ 700010
Additional Details : Weight : 0.05828kg
 View more versions of this product
Product Code. 10613682
  • 133.56 EUR / Each
Estimated Shipment: 08-08-2024
Description
  • Economical mist filters with KF flanges
  • If the filter insert becomes blocked, an integrated pressure control valve opens automatically and reduces the built-up pressure immediately
  • Worn out filter inserts can be replaced easily
Specifications
Specifications
1 Pack
DN 16 KF
Oil Mist Filter