Learn More
Fisherbrand™ Octahedral PTFE Stir Bar
Provides stable stirring action. These PTFE magnetic stir bars are autoclavable and include a pivot ring. The flat surfaces provide a great mixing action.
Brand:  Fisherbrand™
Additional Details : Weight : 0.15480kg
 View more versions of this product
Product Code. 11557782
  • 33.00 EUR / Pack of 10
Estimated Shipment: 18-06-2024
Description
  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action
  • All have pivot rings
  • AlNiCo-V magnets encased in PTFE
  • Autoclavable
Specifications
Specifications
9.5 mm
64 x 9.5mm
10/Pk.
64 mm