Learn More
VACUUBRAND™ NBR O-rings
3.35 EUR - 5.35 EUR
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10224332
|
VACUUBRAND™
23120794 |
71 x 2.5mm |
3.35 EUR
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
11949781
|
VACUUBRAND™
20638139 |
114 x 2mm |
5.35 EUR
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
Specifications
O-Ring | |
NBR | |
1Pack |