Learn More
n-Dodecane, ≥99%, Thermo Scientific Chemicals
Catalog Number p-3345980
Click to view available options
Quantity:
25 g
100 mL
500 mL
2500 mL
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
112-40-3
C12H26
170.34
MFCD00008969
SNRUBQQJIBEYMU-UHFFFAOYSA-N
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
8182
CHEBI:28817
dodecane
CCCCCCCCCCCC
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
112-40-3
C12H26
170.34
MFCD00008969
SNRUBQQJIBEYMU-UHFFFAOYSA-N
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
8182
CHEBI:28817
dodecane
CCCCCCCCCCCC
n-Dodecane is used as a solvent and a distillation chaser. It finds application as a diluent for tributyl phosphate (TBP) in reprocessing plants and as a possible surrogate for kerosene-based fuels in jet. It is an active component of scintillator as well as used in lubricants and greases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Dodecane is used as a solvent and a distillation chaser. It finds application as a diluent for tributyl phosphate (TBP) in reprocessing plants and as a possible surrogate for kerosene-based fuels in jet. It is an active component of scintillator as well as used in lubricants and greases.
Solubility
Miscible with ethyl alcohol, ethyl ether, acetone, chloroform and carbon tetrachloride. Immiscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point °C to 10°C
Density 0.749
Boiling Point 215°C to 217°C
Flash Point 71°C (159°F)
Assay Percent Range ≥99%
Refractive Index 1.422
Quantity 100 mL
Beilstein 1697175
Solubility Information Miscible with ethyl alcohol,ethyl ether,acetone,chloroform and carbon tetrachloride. Immiscible with water.
Formula Weight 170.34
Physical Form Liquid
Chemical Name or Material n-Dodecane