Learn More
WTW™ MultiLine™ Multi 3620 IDS Multi-parameter Portable Meter
Solve multiple measuring tasks at once and save time and costs. WTW™ MultiLine™ Multi 3620 IDS Multi-parameter Portable Meters feature two universal measuring channels for measuring pH, ORP, dissolved oxygen and/or turbidity simultaneously.
Brand: WTW™ 2FD560
1356.96 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Wireless ready
- Simultaneous display of all measured values
- Digital sensor recognition
- Two galvanically isolated measuring inputs
- Dual – 2 channels, 4 parameters
- Galvanic isolation of the sensor inputs prevents interferences of measuring signals
- Use the new wireless modules together with new IDS plug head sensors, be independent from cables and measure even at hardly accessible locations
- Efficient battery or mains supply, you will be able to measure cost-effectively in the field and in the lab
- Data transfer either takes place via USB stick or via an USB connection directly to a computer
- Color graphic display offers measured values as well as additional information such as sensor position, CMC and QSC
- Traceability of the individual measured values can be supported by activating the user administration
Specifications
2 | |
IP 67 | |
Manual 500 / 10000 automatic | |
Wide range power supply, 4x 1.2 V NiMH battery or via Mini USB-B |
LCD Color Graphic, Backlit | |
IDS Multi-parameter Portable Meter only | |
MultiLine™ Multi 3620 IDS Multi-parameter Portable Meter |