Learn More
MP Biomedicals™ Complement, Guinea Pig Purified, 1mL
Brand:  MP Biomedicals™ 0855852
Additional Details : Weight : 0.00100kg
Product Code. 11434235
  • 38.16 EUR / 1mL
Estimated Shipment: 30-07-2024
Specifications
2° to 8°C
Guinea Pig complement
1 mL