missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKRN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12238-1-AP-20UL
This item is not returnable.
View return policy
Description
Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins.Specifications
MKRN2 | |
Polyclonal | |
Unconjugated | |
MKRN2 | |
HSPC070, makorin ring finger protein 2, MKRN2, RING finger protein 62, RNF62 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23609, 297525, 67027 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9ERV1, Q9H000 | |
Mkrn2 | |
MKRN2 Fusion Protein Ag2881 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |