Learn More
Cytiva (formerly Pall Lab) Minimate™ Tangential Flow Filtration (TFF) Capsule with Omega™ Membrane
Concentration and diafiltration (desalting or buffer exchange) processes can be performed on the same system with minimal user intervention.
444.00 EUR - 488.00 EUR
Specifications
Material (Membrane) | PES |
---|---|
Sterility | Non-sterile |
Filtration Area | 50 cm2 |
No. per Pack | 1/Pk. |
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
17164761
|
Cytiva (formerly Pall Lab)
OA001C12 |
1 kDa |
444.00 EUR
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17174761
|
Cytiva (formerly Pall Lab)
OA003C12 |
3 kDa |
444.00 EUR
Each |
Estimated Shipment: 17-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17114771
|
Cytiva (formerly Pall Lab)
OA050C12 |
50 kDa |
444.00 EUR
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17184761
|
Cytiva (formerly Pall Lab)
OA005C12 |
5 kDa |
444.00 EUR
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17134771
|
Cytiva (formerly Pall Lab)
OA100C12 |
100 kDa |
444.00 EUR
Each |
Estimated Shipment: 17-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17144771
|
Cytiva (formerly Pall Lab)
OA300C12 |
300 kDa |
444.00 EUR
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17164771
|
Cytiva (formerly Pall Lab)
OA990C12 |
1000 kDa |
444.00 EUR
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17194761
|
Cytiva (formerly Pall Lab)
OA010C12 |
10 kDa |
444.00 EUR
Each |
Estimated Shipment: 14-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17104771
|
Cytiva (formerly Pall Lab)
OA030C12 |
30 kDa |
488.00 EUR
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17124771
|
Cytiva (formerly Pall Lab)
OA070C12 |
70 kDa |
444.00 EUR
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17154771
|
Cytiva (formerly Pall Lab)
OA500C12 |
500 kDa |
444.00 EUR
Each |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Efficient processing
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
PES | |
Non-sterile | |
50 cm2 | |
1/Pk. |