Learn More
Fisherbrand™ Plan Achromatic Microscope Objective
Use this objective with compound biological microscopes. Fisherbrand™ Plan Achromatic Microscope Objectives are ergonomically designed for everyday use.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Plan achromatic microscope objective
- Accessory for microscope models: FB69075 and FB69088
Specifications
Plan Achromatic Microscope Objective | |
10x/0.25/160/0.17 PH+ spring loaded | |
Microscope models: FB69075 and FB69088 | |
10X |