Learn More
Ansell™ Microgard™ 1800 Standard Coveralls with Hood
Provides comfort in warmer working environments. Ansell Microgard™ 1800 Standard Coveralls with Hood are constructed of lightweight, breathable material yet protects wearer from light spray and dry particles.
308.00 EUR - 385.00 EUR
Specifications
Product Type | Protective Coveralls |
---|---|
Material | Microporous Polyethylene Film |
Color | White |
Waist Style | Elastic |
Ankle Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15516504
|
Ansell™
WH18B-00111-02 |
Small |
385.00 EUR
Pack of 50 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
15526504
|
Ansell™
WH18B-00111-03 |
Medium |
385.00 EUR
Pack of 50 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
15536504
|
Ansell™
WH18B-00111-04 |
Large |
385.00 EUR
Pack of 50 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
15546504
|
Ansell™
WH18B-00111-05 |
X-Large |
385.00 EUR
Pack of 50 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
15556504
|
Ansell™
WH18B-00111-06 |
2X-Large |
308.00 EUR
Pack of 40 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
15566504
|
Ansell™
WH18B-00111-07 |
3X-Large |
308.00 EUR
Pack of 40 |
Estimated Shipment: 20-11-2024
Log in to see stock. |
|||||
Description
- Coverall with 3-piece hood
- Elasticated wrists, waist and ankles
- 2-way front zipper with re-sealable storm flap
- Bound seams
- Standard: CE category III, type 5/6 EN 1149-5 (antistatic)
- Low hazard liquid spray, splash and particle protection
- Moisture vapour permeable (‘breathable’) to help reduce the risk of heat stress
- Silicone-free which is critical in spray painting applications
- Anti-static according to EN1149-5
- Low linting to reduce the risk of fibre contamination in some critical areas
- Optimised body fit for wearer comfort and safety
Specifications
Protective Coveralls | |
White | |
Elastic | |
Hooded | |
2-way Front Zipper with Storm Flap | |
CE category III, type 5/6 EN 1149-5 | |
Includes Hood |
Microporous Polyethylene Film | |
Elastic | |
Elastic | |
3-panel | |
Bound | |
None |