Learn More
Metro™ MetroMax Shelf Posts
For use with Metro™ MetroMax i™ and Q™ shelving
Brand:  Metro™ MX54P
Additional Details : Weight : 0.72800kg
Product Code. 15927211
  • 58.85 EUR / Each
Estimated Shipment: 16-09-2024
Description
Grooves located every 1 in. for locking shelf wedges in place. Post height stamped to face of post for easy shelf positioning.
  • Corrosion-proof polymer posts with built-in Microban™ antimicrobial product protection
  • Stationary posts include an adjustable leveling foot to compensate for uneven floors
  • Leveling foot can be adjusted 1 in. (25mm)
  • Mobile posts accept optional Metro Stem Casters, available in a variety of materials to meet your specific needs
Specifications
Specifications
Stationary Post with Leveling Foot
54 in.
Polymer
137.1 cm
Metro™ MetroMax i™ and Q™ shelving