missing translation for 'onlineSavingsMsg'
Learn More
Learn More
γ-Methacryloxypropyltrimethoxysilane, 99.04%, MP Biomedicals™
Click to view available options
Quantity:
5 mL
25 mL
Specifications
Color | Colorless |
Density | 1.04g/mL |
Quantity | 5 mL |
Formula Weight | 248.35 |
Percent Purity | 0.9904 |
Physical Form | Liquid |
Chemical Name or Material | γ-Methacryloxypropyl-trimethoxysilane, Adhesive |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'