Learn More
Lovibond™ MC500, MD600 Colorimeter Sample Cells
For use with MC500, MD600 colorimeters
Brand:  Lovibond™ 197620
Additional Details : Weight : 0.22000kg
 View more versions of this product
Product Code. 12747837
  • 88.60 EUR / Pack of 12
Estimated Shipment: 16-07-2024
Description
For use with MC500, MD600 colorimeters
  • Material: glass
  • Volume: 10mL
  • With lids
Specifications
Specifications
24 mm
SP600 Spectrophotometer
Glass
Colorimeter Sample Cells
12 Pk. of sample cells with lids and light shielding rings
10 mL