Learn More
Lovibond™ Aqua Comparitor Color Discs: Copper
For testing water with the Aqua Comparator
26.10 EUR
Specifications
For Use With (Application) | Testing water with the Aqua comparator™ |
---|---|
For Use With (Equipment) | Aqua comparator™ |
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
10631304
|
Lovibond™
146430 |
Copper HR Color Disc | 0 to 5mg/L |
26.10 EUR
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
11745528
|
Lovibond™
146440 |
Copper LR Color Disc | 0 to 1mg/L |
26.10 EUR
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for copper using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ |