Learn More
UVP LED White Light Plate for UVP Imagers
For colorimetric imaging in the darkroom via uniform, backlit white light
Brand: UVP 95-0476-01
Additional Details : Weight : 3.00000kg
Description
- View autoradiographs, as well as Coomassie Blue and Silver stains
- Dimensions: 11W x 11D in. (28 x 28cm)
- For use with UVP™ Imagers
Specifications
White | |
For use with uvp™ imagers | |
For colorimetric imaging in the darkroom via uniform, backlit white light | |
Uv Light Plate |