Learn More
Cytiva (formerly Pall Lab) Laboratory Manifold Elongated Adapter for Number 8 Stopper
Accessory elongated adapter for microbiology manifold.
Brand:  Cytiva (formerly Pall Lab) 4959
503.50 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
Product Code. 17174451
  • 530.00 EUR / Pack of 3
Estimated Shipment: 06-11-2024
Description
  • Lab manifold is designed for flexibility and reduction in cross-contamination
  • Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black Fluorocarbon Elastomer O-ring
Stainless Steel (316L)
3/Pk.
Number 8 Stopper
Non-sterile
Elongated