Learn More
DWK Life Sciences Kimble™ Kontes™ Clamps
For 90mm filter
330.00 EUR - 1330.00 EUR
Specifications
For Use With (Equipment) | All-glass ultra-ware microfiltration apparatus with stainless steel support |
---|---|
Diameter (Metric) Inside | 90 mm |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11843123
|
DWK Life Sciences
953753-0090 |
Aluminum Clamp |
330.00 EUR
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
11883123
|
DWK Life Sciences
953841-0090 |
Support Base |
1330.00 EUR
Each |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
Description
- Component part for all-glass ultra-ware microfiltration apparatus with stainless steel support
Specifications
All-glass ultra-ware microfiltration apparatus with stainless steel support | |
90 mm |