Learn More
Ismatec™ BVP-Z Gear Pump Drive
Use this drive for pulseless fluid delivery. Ismatec™ BVP-Z Gear Pump Drives feature closed-loop control for pressure independent speed.
Brand: Ismatec™ ISM446B
This item is not returnable.
View return policy
Description
- Powerful, variable speed delivery pump
- Digital potentiometer for speed setting
- 20 interchangeable pump-heads
- Analog interface
Specifications
LCD | |
50/60 Hz | |
BVP-Z | |
220 V |
220 V | |
Foot switch | |
Pump Drive |