missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) Chloride Hexahydrate, ≥98%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Boiling Point | 280°C to 285°C |
Melting Point | 37°C |
Color | Brown |
pH | 1.8 (at 10.00g/L 25°C) |
Physical Form | Solid |
Quantity | 250 g |
Linear Formula | FeCl3 · 6H2O |
UN Number | UN3260 |
Formula Weight | 270.3g/mol |
Percent Purity | ≥98% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'