Learn More
INTS3 Mouse anti-Human, Unconjugated, Clone: 2G11B7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67060-1-IG-150UL
This item is not returnable.
View return policy
Description
The INTS3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human INTS3. This antibody recognizes human, mouse, rat antigen. The INTS3 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
INTS3 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunofluorescence, ELISA | |
2G11B7 | |
Unconjugated | |
150 μL | |
65123 | |
IgG2b |