Learn More
IKA Works MF10 Micro Mill Sieves
Brand:  IKA 0002939200
Additional Details : Weight : 1.00000kg
Product Code. 10573831
  • 399.00 EUR / Each
Estimated Shipment: 23-07-2024
Specifications
Stainless Steel
1Each
Sieve
1 mm
1.0mm