missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hIST1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | hIST1 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16819324
|
Proteintech
19842-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16809324
|
Proteintech
19842-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.Specifications
hIST1 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P53990, Q568Z6, Q9CX00 | |
307833, 71955, 9798 | |
hIST1 Fusion Protein Ag13874 | |
Primary | |
-20°C | |
IST1 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
hIST1, IST1, IST1 homolog, KIAA0174, OLC1 | |
Ist1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |