Learn More
Karl Hecht™ Hirsch Funnel
Filter materials through this funnel using filter paper and vacuum suction. Karl Hecht™ Hirsch Funnel has sloped sides.
Brand:  Karl Hecht™ 7006/70
Additional Details : Weight : 0.06000kg
Product Code. 11327464
  • 99.00 EUR / Pack of 10
Estimated Shipment: 19-06-2024
Description
  • Filtering funnel.
  • Sloped sides.
  • Efficient and easy filtration.
Specifications
Specifications
Funnel
28mm
Filter paper, 28mm diameter