Learn More
Thermo Scientific™ Herasafe™ KS Options and Accessories
Worksurfaces, exhaust transitions and baskets
Brand: Thermo Scientific™ 50078796
122.83 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Worksurfaces, exhaust transitions and baskets
Specifications
Thimble Duct Exhaust Transition for Herasafe KS 9 (3 ft) Cabinet | |
Thimble Duct Exhaust Transition Accessory | |
Biosafety Cabinet |