Learn More
Liebherr™ Heavy-Duty Laboratory Refrigerator
Brand: Liebherr™ LKPv 1420
Additional Details : Weight : 199.00000kg
Description
A range of purpose built laboratory refrigerators, single and double door, solid and glass door.- Data logging of temperature every two minutes
- Audible and visual door-open and hi-low temperature alarm
- Volt free contact for connection to remote system
- Data transferable to external unit