Learn More
Brand™ Wide-mouth Square HDPE Bottles
Manufactured from translucent HDPE. BRAND™ Wide-mouth Square HDPE Bottles save space on tight counters, benches, and cabinets. Tamper-proof polypropylene closure includes sealing cone with press-off retaining ring. Ideal for use as a storage container.
1.74 EUR - 10.35 EUR
Specifications
Color | Translucent |
---|---|
Includes | Press-off retaining ring |
Material | HDPE |
Closure Color | Blue |
Closure Included | Yes |
Product Code | Brand | Capacity (Metric) | Thread Size (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Thread Size (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | |||
11613129
|
Brand™
130412 |
100 mL | GL 32 | 45 x 45 x 101mm | |||||
11623129
|
Brand™
130414 |
250 mL | GL 45 | 60 x 60 x 127mm | |||||
11633129
|
Brand™
130418 |
500 mL | GL 54 | 75 x 75 x 161mm | |||||
11643129
|
Brand™
130422 |
1000 mL | GL 54 | 85 x 85 210mm | |||||
11653129
|
Brand™
130424 |
1500 mL | GL 80 | 111 x 111 x 209mm | |||||
10121871
|
Brand™
130426 |
2500 mL | GL 80 | 122 x 122 x 258mm | |||||
11663129
|
Brand™
130428 |
4000 mL | GL 80 | 142 x 142 x 294mm | |||||
Description
- Manufactured from translucent HDPE.
- Tamper-proof polypropylene closure includes sealing cone with press-off retaining ring.
- Rectangular shape saves space on tight counters, benches, and cabinets.
- Ideal for use as a storage container.
Specifications
Translucent | |
HDPE | |
Yes | |
Square | |
Polypropylene |
Press-off retaining ring | |
Blue | |
Wide | |
Square Bottle | |
Screw Cap with Sealing Cone |