Learn More
Thermo Scientific Chemicals Graphite Gas Analysis Crucible; Ht (mm), 24.5; ID (mm), 10.0; OD (mm), 12.7; Tip Dia (mm), 6.80
103.00 EUR - 154.00 EUR
Specifications
Material | Graphite |
---|---|
For Use With (Application) | For Gas analysis |
Product Type Specs | Graphite Crucible |
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | ||||
15473705
|
Thermo Scientific Chemicals
039809.KN |
12.6mm | 24.2mm | ||||||
15411577
|
Thermo Scientific Chemicals
039811.KN |
12.7mm | 24.5mm |
154.00 EUR
Pack of 100 |
Estimated Shipment: 28-10-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Graphite | |
For Gas analysis | |
Graphite Crucible |
RUO – Research Use Only