Learn More
Fisherbrand™ Grade 1103 Cellulose Technical Filter Paper
Suitable for removal of fine particles
Brand: Fisherbrand™
Additional Details : Weight : 0.31500kg
Description
- Thick smooth paper
- Wet strengthened
Specifications
Disc | |
12 to 16 μm | |
For dyes, Ether, Alcoholic solutions, Eaux de Cologne, salted solutions, vegetal extracts, hair solutions, perfumes, tinctures, saccharine juices, vinegar | |
100 Pack | |
150 mm |
Cellulose | |
12 to 16 μm | |
1103 | |
0.3 mm | |
150 mm |