Learn More
Sartorius Stedim Biotech™ Glass Microfiber Filters, Grade 1336/2
Generally used for air and gas monitoring, but are suitable as a prefilter.
Brand: Sartorius Stedim Biotech™ FT-201120-210297
Additional Details : Weight : 0.45600kg
Description
- Produced with synthetic binders to make them more resistant
- Suitable for air monitoring or prefiltration
- Non-sterile, bulk packed
Specifications
Sheet | |
Non-sterile | |
Hydrophobic | |
0.4 mm |
Glass Microfiber with Binder | |
1336/2 | |
100/Pk. | |
210 x 297 mm |