Learn More
Gilson™ Accessory for PIPETMAN™ Complete Pipetting System- Coloris™ Colored Identifi cation Clips
GLP-friendly, brightly colored clips made of polyacetal are used for identifying Pipetteman™ and Neo™ pipets
Brand: Gilson™ F161306
Additional Details : Weight : 0.50000kg
Description
- Brightly colored clips
- Made of polyacetal
- Used for identifying Pipetteman and Neo pipettes
Specifications
White | |
Polyacetal | |
Pipetteman™ and Neo™ pipettes |