Learn More
Epredia™ Gill™ Hematoxylin 1, 2, and 3
With three strengths available, the Epredia™ Gill™ 1, 2, 3 Hematoxylin allows complete control of staining intensities.
94.65 EUR - 192.00 EUR
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | ||||
10624009
|
Epredia™
72404 |
Gill 1 Hematoxylin | 1 pt. | ||||||
11326740
|
Epredia™
72411 |
Gill 1 Hematoxylin | 1 gal. |
192.00 EUR
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||||
10289688
|
Epredia™
72504 |
Gill 2 Hematoxylin | 1 pt. | ||||||
10444939
|
Epredia™
72511 |
Gill 2 Hematoxylin | 1 gal. |
152.00 EUR
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||||
10580139
|
Epredia™
72604 |
Gill 3 Hematoxylin | 1 pt. | ||||||
10590139
|
Epredia™
72611 |
Gill 3 Hematoxylin | 1 gal. |
147.00 EUR
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||||
Description
Gill Hematoxylins are progressive nuclear stains.- Gill 1 is recommended for use in cytology
- Gill 2 has applications in histology and cytology
- Gill 3 is used in histology
- All are pre-mixed and ready-to-use
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H371-May cause damage to organs.
Precautionary Statement
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : aluminium sulphate octadecahydrate, sodium iodate, hematoxylin, ethylene glycol,diazolidinyl urea,